missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BCR-ABL Monoclonal Antibody (7C6), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA1153
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Western blot analysis of MA1-153 detects ∽130-160 kDa and a 210-250 kDa protein in K562 cells. In addition to BCR-ABL, BCR and ABL are also expressed in these cells and full length BCR is also detected by the antibody. For detection purposes, non-reducing conditions resulted in enhanced detection of BCR-ABL.
A reciprocal translocation between chromosomes 22 and 9 produces the BCR-ABL fusion oncoprotein, which is found in patients with chronic myelogenous leukemia (CML). Although there are various versions, p210 is the most commonly found BCR-ABL form. The fusion to BCR, results in deregulated/constitutively active tyrosine kinase activity of BCR-ABL which is sufficient of oncogenic transformation of myeloid cells. Inhibition of BCR-ABL oncogenic activity by the ABL-kinase inhibitor STI-571 (also known as gleevec, imatinib) became the first and best example for successful targeted therapy of cancer, and specifically CML. In recent years, several mutations (such as the T351I mutation) were found in CML patients that resulted in STI-571 resistance and the need for additional drug development.
Especificaciones
BCR-ABL | |
Monoclonal | |
1 mg/mL | |
PBS with 1mg/mL BSA and 0.05% sodium azide | |
P11274, Q6PAJ1 | |
BCR | |
Bcr686 thyroglobulin conjugate corresponding to human BCR sequence 686-696 (SSINEEITPRRQS) | |
100 μg | |
Primary | |
Human, Mouse | |
Antibody | |
IgG2a |
Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
7C6 | |
Unconjugated | |
BCR | |
5133400C09Rik; AI561783; AI853148; ALL; BCR; BCR, RhoGEF and GTPase activating protein; BCR/FGFR1 chimera protein; BCR1; breakpoint cluster region; breakpoint cluster region homolog; breakpoint cluster region protein; CML; D22S11; D22S662; FGFR1/BCR chimera protein; Kiaa3017; mKIAA3017; PHL; Renal carcinoma antigen NY-REN-26 | |
Mouse | |
Protein A | |
RUO | |
110279, 613 | |
-20°C | |
Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
BCR-ABL Monoclonal Antibody (7C6), Invitrogen™ > 100 μg; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido