Learn More
BD Difco™ & BBL™ Manual (Second Edition)
The Manual of Microbiological Culture Media is a valuable resource for all laboratories using dehydrated, bottled, tubed and plated media. BD Difco™ & BBL™ Manual (Second Edition) includes a CD with an electronic copy for easy access and searching.
Marca:  BD 220225
Código de producto. 11708313
  • 94.85€ / cada uno
Fecha estimada de envío: 28-04-2025
para ver el stock
Explore más ofertas especiales
Descripción
This second edition combines the Difco™ Manual (11th edition) and the Manual of BBL™ Products and Laboratory Procedures (6th edition). Manual has the entire line of Difco and BBL media including dehydrated culture media (DCM) and prepared plated, tubed, and bottled media.
Product descriptions include
  • Medium formulation
  • Recommended quality control organisms and expected results
  • History, ingredients, and usage information
  • Catalog numbers for all packaging configurations
  • Icons denoting “official” and “standard methods” publications
Also includes flowcharts, application tables, over 400 color photographs, and CD-ROM of manual.
TRUSTED_SUSTAINABILITY
Especificaciones
Especificaciones
For laboratory use
Certificados
Ofertas especiales
Ofertas especiales
Título del producto
BD Difco™ & BBL™ Manual (Second Edition) > Difco and BBL Manual; Second edition