Learn More
BD Slip Tip Sterile Syringes
Slip-tip disposable syringe
Especificaciones
Certificaciones/conformidad OSHA
Para utilizar con (aplicación) General purpose
Sin látex Sí
Tipo de producto Jeringa de tuberculina
Desechables Single-use
Ver más especificaciones
Descripción
Descripción
Slip-tip disposable syringe
  • Clear barrel with bold scale markings
  • Tapered plunger rod for ease of aspiration and a positive plunger rod stop
  • Labeled packaging
  • Latex-free
Especificaciones
Especificaciones
OSHA
Sí
Single-use
Sí
General purpose
Jeringa de tuberculina
Slip Tip
Estéril
Documentos
Product Certifications
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.