missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beclin 1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 46 publications
Marca: Novus Biologicals NB110-87318
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Beclin 1 Polyclonal specifically detects Beclin 1 in Human, Mouse, Rat, Porcine, Alligator, Canine, Primate samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Knockdown Validated.
Especificaciones
Beclin 1 | |
Polyclonal | |
Unconjugated | |
PBS, 0.02 Sodium Azide with 0.02% Sodium Azide | |
ATG6, ATG6 autophagy related 6 homolog, beclin 1 (coiled-coil, moesin-like BCL2 interacting protein), beclin 1 (coiled-coil, moesin-like BCL2-interacting protein), beclin 1, autophagy related, beclin1, beclin-1, Coiled-coil myosin-like BCL2-interacting protein, GT197, Protein GT197, VPS30 | |
Rabbit | |
51 kDa | |
0.05 mL | |
Apoptosis, Autophagy, Cancer, HIF Target Genes, Hypoxia, Immunology, Innate Immunity, mTOR Pathway, Tumor Suppressors | |
8678 | |
Human, Mouse, Rat, Pig, Alligator, Canine, Primate | |
IgG |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), KnockDown | |
1 mg/mL | |
Western Blot 1:1000, Simple Western 1:12.5, Immunohistochemistry 1:50 - 1:400, Immunocytochemistry/Immunofluorescence 1:50, Immunohistochemistry-Paraffin 1:50 - 1:400, KnockDown Validated reported in scientific literature (PMID 29297744) | |
Q14457 | |
BECN1 | |
Synthetic peptide made to an internal portion of human Beclin 1 (within residues 150-300). [Swiss-Prot: Q14457] | |
Affinity Purified | |
RUO | |
Primary | |
Human, mouse, rat (96%), monkey, bovine (97%), porcine. Canine reactivity reported in scientific literature (PMID: 24027311). Possible reactivity to Chicken (87%), Xenopus laevis (79%), and Xenopus tropicalis (77%) (not yet validated). | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Beclin 1 Antibody - BSA Free, Novus Biologicals™ > 0.05mL; Unlabeled
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido