missing translation for 'onlineSavingsMsg'
Learn More
Learn More
behr Labor-Technik™ behropur™ Water Deionizer For Depressurised Operation
Thick-walled, robust and practical mixed-bed ion exchangers made of blue polyethylene with free output to the storage tank. Ideal also for subsequent demineralization of reverse osmosis system or for an ambitious aquarist.
505.00€ - 960.00€
Código de producto | Marca | Modelo | Capacidad | Precio | Cantidad y disponibilidad | ||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Modelo | Capacidad | Precio | Cantidad y disponibilidad | ||||
10069090
|
behr Labor-Technik™
B00223465 |
B5Z | 500 l |
505.00€
cada uno |
Fecha estimada de envío: 12-09-2024
Inicie sesión para ver el stock |
||||
10099110
|
behr Labor-Technik™
B00223466 |
B10Z | 1000 l |
960.00€
cada uno |
Fecha estimada de envío: 12-09-2024
Inicie sesión para ver el stock |
||||
Descripción
- Can be connected directly to the water supply and is immediately ready for use
- Automatic deaeration due to the water being supplied from the bottom
- Minimum risk of microbial contamination due to slit nozzles in the raw water supply
- Nozzles are sturdy and easy to clean, no risk of damage due to mechanical stress
- Extremely secure and leakproof sealing of the nozzles due to the manufacturer’s own welding process
- Conductivity meter is mounted directly on top
- B5 and B10 can also be used as wall units (mounting brackets included in the scope of supply)
- Also available with shutoff when the limit value is reached and level control
- Conductance is controlled directly on the unit (installed measuring equipment) or with remote location meter
- Conductance is controlled directly on the unit (installed measuring equipment) or with remote location meter
- Also available with shutoff when the limit value is reached and level control
- B5 and B10 can also be used as wall units (mounting brackets included in the scope of supply)
- Conductivity meter is mounted directly on top
- Extremely secure and leakproof sealing of the nozzles due to the manufacturer’s own welding process
- Nozzles are sturdy and easy to clean, no risk of damage due to mechanical stress
- Minimum risk of microbial contamination due to slit nozzles in the raw water supply
- Automatic deaeration due to the water being supplied from the bottom
Especificaciones
Polietileno | |
Cartucho de repuesto |