Learn More
Invitrogen™ beta-2 Defensin Human ELISA Development Kit (ABTS), PeproTech®, PeproTech®
ELISA
Marca: Invitrogen™ 900-M172
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Defensins (alpha and beta) are cationic peptides with a broad spectrum of antimicrobial activity that comprise an important arm of the innate immune system. The alpha-defensins are distinguished from the beta-defensins by the pairing of their three disulfide bonds. To date, six human beta-defensins have been identified; BD-1, BD-2, BD-3, BD-4, BD-5 and BD-6. beta-defensins are expressed on some leukocytes and at epithelial surfaces. In addition to their direct antimicrobial activities, they can act as chemoattractants towards immature dendritic cells and memory T cells. The beta-defensin proteins are expressed as the C-terminal portion of precursors, and are released by proteolytic cleavage of a signal sequence and, in some cases, a propeptide sequence. Beta-defensins contain a six-cysteine motif that forms three intra-molecular disulfide bonds.Especificaciones
O15263 | |
1673 | |
2 x 96 pruebas | |
Refer to product documentation for component specific storage temperature. |
ELISA | |
DEFB4A | |
RUO | |
Humano |