missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta 2-Microglobulin Antibody (BBM.1), Alexa Fluor™ 350, Novus Biologicals™
Rabbit Monoclonal Antibody
Marca: Novus Biologicals NBP2-81045AF350
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
beta 2-Microglobulin Monoclonal specifically detects beta 2-Microglobulin in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Especificaciones
| beta 2-Microglobulin | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50mM Sodium Borate | |
| B2M | |
| Molt 4, a human T cell line. | |
| 0.1 mL | |
| Adaptive Immunity, Cancer, Cell Biology, Diabetes Research, Immunology | |
| 567 | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| BBM.1 | |
| Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| beta chain of MHC class I molecules, beta-2-microglobin, beta-2-microglobulin | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Binds human beta 2-Microglobulin, a component of MHC class I molecules, which are present on all nucleated cells (excludes red blood cells) and involved in the presentation of peptide antigens to the immune system. | |
| Store at 4°C in the dark. | |
| IgG κ |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido