Learn More
beta-5 Defensin Polyclonal Antibody, PeproTech®, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen 500-P323-50UG
Descripción
AA Sequence of recombinant protein: GLDFSQPFPS GEFAVCESCK LGRGKCRKEC LENEKPDGNC RLNFLCCRQR I. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Human BD-5. Anti-Human BD-5-specific antibody was purified by affinity chromatography employing an immobilized Human BD-5 matrix. Sandwich ELISA: To detect Human BD-5 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.5-2.0 μg/mL of this antibody is required. This antigen affinity purified antibody, in conjunction with PeproTech Biotinylated Anti-Human BD-5 (500-P323Bt) as a detection antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant Human BD-5. Western Blot: To detect hBD-5 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant hBD-5 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions.
Defensins (alpha and beta) are cationic peptides with a broad spectrum of antimicrobial activity that comprise an important arm of the innate immune system. The alpha-defensins are distinguished from the beta-defensins by the pairing of their three disulfide bonds. To date, six human beta-defensins have been identified; BD-1, BD-2, BD-3, BD-4, BD-5 and BD-6. beta-defensins are expressed on some leukocytes and at epithelial surfaces. In addition to their direct antimicrobial activities, they can act as chemoattractants towards immature dendritic cells and memory T cells. The beta-defensin proteins are expressed as the C-terminal portion of precursors, and are released by proteolytic cleavage of a signal sequence and, in some cases, a propeptide sequence. Beta-defensins contain a six-cysteine motif that forms three intra-molecular disulfide bonds.Especificaciones
beta-5 Defensin | |
Polyclonal | |
Unconjugated | |
DEFB105A | |
DEFB105A, DEFB105B | |
E.coli-derived Recombinant Human BD-5 | |
50μg | |
Primary | |
Human | |
Lyophilized |
ELISA, Western Blot | |
0.1-1.0 mg/mL | |
PBS with no preservative | |
Q8NG35 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
245908, 504180 | |
-20°C |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.