missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beta-catenin Mouse anti-Canine, Human, Clone: OTI2H3, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Marca: Origene Technologies CF500049
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Beta-catenin, an adherens junction (AJ) protein, was originally identified as a component of cell-cell adhesion structures. AJs are necessary for the creation and maintece of epithelial cell layers by regulating cell growth and adhesion between cells. Beta-catenin interacts with the cytoplasmic domain of E-cadherin and links E-cadherin to alpha-catenin, which in turn mediates anchorage of the E-cadherin complex to the cortical actin cytoskeleton. Studies show that Beta-catenin also binds to another cytoskeletal complex containing the adenomatous polyposis coli protein and microtubules, and interacts with several signaling pathways that include tyrosine kinases, phosphatases and Wnt/Wingless. The interplay between beta-catenin, cytoskeletal complexes and signaling pathways may regulate morphogenesis. Beta-catenin is expressed in several hair follicle cell types, basal and peripheral matrix cells, and cells of the outer and inner root sheats. A pathological role of beta-catenin has been identified in pilomatrixoma (PTR), medulloblastoma (MDB), colorectal cancer (CRC), ovarian cancer, and tumor development. In the nucleus, beta-catenin serves to co activate a family of Lef/Tcf transcription factors that stimulate transcription of target genes including those encoding cyclin D and c-myc that promote cell proliferation. The influence on cell proliferation is the molecular basis for the role of beta-catenin in tumorgenesis, specifically, solid tumors of the breast, colon, liver, lung, gastric, prostate, and skin.Especificaciones
Beta-catenin | |
Monoclonal | |
Unconjugated | |
CTNNB1 | |
CTNNB, EVR7, MRD19, armadillo | |
Mouse | |
Affinity Chromatography | |
RUO | |
1499, 477032 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI2H3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
B6V8E6, P35222 | |
CTNNB1 | |
Human recombit protein fragment corresponding to amino acids 531-781 of human beta-catenin produced in E.coli. | |
100 μg | |
Primary | |
Canine, Human | |
Antibody | |
IgG2a |