missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beta-Tubulin III Mouse, anti-Human, Unconjugated, Clone: AA10, STEMCELL Technologies
Descripción
- The AA10 antibody reacts with beta-tubulin III, an ∼50 - 55 kDa structural protein that is a constituent of tubulin
- Tubulin is the major component of microtubules within the cytoskeleton and is assembled from heterodimers of alpha and beta tubulin subunits
- The beta III isoform of tubulin, also known as neuron-specific class III beta-tubulin, is expressed primarily in neurons and is widely used as a marker to distinguish neurons from other cell types
- Beta-tubulin III contributes to microtubule formation in neuronal cell bodies and axons, a function involving GTP binding, and plays roles in axonal transport, neuronal cell proliferation, and differentiation
- It is highly expressed in several types of cancer and is a predictive and prognostic marker for various tumors, for example, being found in neoplastic but not in normal glial cells
- The AA10 antibody is expected to recognize all mammalian homologs of beta-tubulin III
Especificaciones
Especificaciones
| Antígeno | Beta-Tubulin III |
| Aplicaciones | Immunohistochemistry, Western Blot, Flow Cytometry, Immunocytochemistry, Immunofluorescence |
| Clasificación | Monoclonal |
| Clon | AA10 |
| Conjugado | Unconjugated |
| Especie del huésped | Mouse |
| Cantidad | 25 μg |
| Primario o secundario | Primary |
| Especies diana | Human, Rat, Mouse |
| Isotype | IgG2a κ |
| Mostrar más |
Título del producto
Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.
¿Detecta una oportunidad de mejora?