missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bif1 Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marca: Invitrogen PA19017
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
This antibody is predicted to react with bovine, canine, mouse and rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
Bif1 gene encodes a protein similar to a protein in rodents which is induced by bone morphogenic protein 2 in vitro and may be involved in BMP2-induced transcription.
Especificaciones
Bif1 | |
Polyclonal | |
Unconjugated | |
Sh3glb1 | |
AA409932; AI314629; AU015566; Bax-interacting factor 1; BIF1; Bif-1; CGI-61; dJ612B15.2; EMBL:AAH79085.1}; endophilin b1; endophilin-B1; Kiaa0491; mKIAA0491; PPP1R70; protein phosphatase 1, regulatory subunit 70; SH3 domain containing GRB2 like endophilin B1; SH3 domain-containing GRB2-like protein B1; SH3-containing protein SH3GLB1; SH3-domain GRB2 like endophilin B1; SH3-domain GRB2-like B1 (endophilin); SH3-domain GRB2-like endophilin B1; SH3GLB1; sh3glb1 {ECO:0000312; testicular tissue protein Li 172 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
292156, 51100, 54673 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q6AYE2, Q9JK48, Q9Y371 | |
Sh3glb1 | |
Synthetic peptide corresponding to the C terminus amino acids QKGKVPITYLELLN. | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Bif1 Polyclonal Antibody, Invitrogen™ > 100 μg; Unconjugated
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido