missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ BIK (Human) Recombinant Protein
Human BIK full-length ORF ( AAH01599, 1 a.a. - 160 a.a.) recombinant protein with GST-tag at N-terminal.
Marca: Abnova™ H00000638-P01.10ug
Detalles adicionales : Peso : 0.00010kg
Descripción
- Sequence: MSEVRPLSRDILMETLLYEQLLEPPTMEVLGMTDSEEDLDPMEDFDSLECMEGSDALALRLACIGDEMDVSLRAPRLAQLSEVAMHSLGLAFIYDQTEDIRDVLRSFMDGFTTLKENIMRFWRSPNPGSWVSCEQVLLALLLLLALLLPLLSGGLHLLLK
Especificaciones
AAH01599 | |
BCL2-interacting killer (apoptosis-inducing) | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
BIK | |
GST |
638 | |
Wheat germ expression system | |
10 μg | |
BIP1, BP4, NBK | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |