Learn More
ACROBiosystems Biotinylated Human B7-H5 / Gi24 / VISTA Protein, Fc, Avitag™
Biotinylated Protein
Marca: ACROBiosystems B75-H82F3-200UG
Detalles adicionales : Peso : 0.00001kg
Descripción
Platelet receptor Gi24, also known as B7-H5 and stress-induced secreted protein-1 (Sisp-1), is a protein that in humans is encoded by the C10orf54 gene, which contains 1 Ig-like (immunoglobulin-like) domain. As for C10orf54 gene, C10orf54 appears to positively interact with BMP-4, potentiating BMP signaling and the transition from an undifferentiated to a differentiated state on ESCs. Human C10orf54 undergoes proteolytic cleavage by MT1-MMP, generating a soluble 30 kDa extracellular fragment plus a 25-30 kDa membrane-bound fragment.
This protein carries a human IgG1 Fc tag at the C-terminus, followed by a Avi tag (Avitag™). The protein has a calculated MW of 47.1 kDa. The protein migrates as 55-66 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
AAH20568.1 | |
200 μg | |
Phe 33 - Ala 194 | |
B7-H5,SISP1,Gi24,VISTA | |
Precision Avi | |
Fc Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
47.1 kDa | |
Biotinylated Human B7-H5, Fc,Avitag (B75-H82F3) is expressed from human 293 cells (HEK293). It contains AA Phe 33 - Ala 194 (Accession # AAH20568.1). | |
-20°C | |
B7-H5 | |
Human | |
Tris with Glycine, Arginine and NaCl, pH7.5 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |