missing translation for 'onlineSavingsMsg'
Learn More

ACROBiosystems Biotinylated Human Fc gamma RIIIA / CD16a (F176) Protein, Avitag™,His Tag (SPR & BLI verified)

Biotinylated Protein

Marca:  ACROBiosystems CDA-H82E8-25UG

Detalles adicionales : Peso : 0.00001kg

 Ver más versiones de este producto

Código de producto. 15876347

  • 302.42€ / 25 microgramos
Fecha estimada de envío: 07-08-2024
para ver el stock

Descripción

Descripción

CD16 is a low affinity Fc receptor, and has been identified as Fc receptors FcγRIIIa (CD16a) and FcγRIIIb (CD16b). These receptors bind to the Fc portion of IgG antibodies. CD16 encoded by two different highly homologous genes in a cell type-specific manner.CD16 is found on the surface of natural killer cells, neutrophil polymorphonuclear leukocytes, monocytes and macrophages.CD16a antigen is also known as Low affinity immunoglobulin gamma Fc region receptor III-A, Fc-gamma RIII-alpha. CD16b is a low-affinity, GPI-linked receptor expressed by neutrophils and eosinophils, whereas CD16a is an intermediate affinity polypeptide-anchored transmembrane glycoprotein expressed natural killer cells, macrophages, subpopulation of T-cells, immature thymocytes and placentaltrophoblasts.CD16a is involved in phagocytosis, secretion of enzymes and inflammatory mediators, antibody­dependent cytotoxicity and clearance of immune complexes. Aberrant expression or mutations of CD16a is implicated in susceptibility to recurrent viral infections, systemic lupus erythematosus, and alloimmune neonatal neutropenia.

This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 25.5 kDa. The protein migrates as 40-55 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.
Especificaciones

Especificaciones

25.5 kDa
Biotinylated Human CD16a (F176), Avitag,His Tag (SPR and BLI verified) (CDA-H82E8) is expressed from human 293 cells (HEK293). It contains AA Gly 17 - Gln 208 (Accession # P08637-1).
-20°C
Fc gamma RIIIA / CD16a
Human
PBS, pH7.4
HEK293
MABSol™ Biotin Labeled Proteins
25 μg
Gly 17 - Gln 208
FCGR3A,CD16A,FCG3,FCGR3,IGFR3
Precision Avi
His Tag and Avi Tag
Less than 1.0 EU / μg by the LAL method.
Powder
>95% as determined by SDS-PAGE
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales