Learn More
ACROBiosystems Biotinylated Human G-CSF R / CD114 Protein, Avitag™,His Tag
Biotinylated Protein
Marca: ACROBiosystems GCR-H82E4-25UG
Detalles adicionales : Peso : 0.00001kg
Descripción
The granulocyte colony-stimulating factor receptor (G-CSF-R) also known as CD114 (Cluster of Differentiation 114) is a protein that is encoded by the CSF3R gene. G-CSF-R is a cell-surface receptor for the granulocyte colony-stimulating factor (G-CSF). The G-CSF receptors belongs to a family of cytokine receptors known as the hematopoietin receptor family. Granulocyte colony-stimulating factor receptor has been shown to interact with Grb2, HCK and SHC1. The granulocyte colony-stimulating factor receptor is present on precursor cells in the bone marrow, and, in response to stimulation by G-CSF, initiates cell proliferation and differentiation into mature neutrophilic granulocytes and macrophages.
This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 69.2 kDa. The protein migrates as 72-85 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Especificaciones
NP_000751 | |
25 μg | |
Glu 25 - Pro 621 | |
CSF3R,CD114,GCSFR | |
Precision Avi | |
His Tag and Avi Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
69.2 kDa | |
Biotinylated Human G-CSF R, Avitag,His Tag (GCR-H82E4) is expressed from human 293 cells (HEK293). It contains AA Glu 25 - Pro 621 (Accession # NP_000751). | |
-20°C | |
G-CSF R | |
Human | |
PBS, pH7.4 | |
HEK293 | |
MABSol™ Biotin Labeled Proteins |