missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Propionato de acetato de bisfenol A, epoxi curativa, CE-326, Thermo Scientific™
41.05€ - 116.00€
Identificadores de productos químicos
CAS | 1071466-57-3 |
---|---|
Fórmula molecular | C20H22O4 |
Peso molecular (g/mol) | 326.392 |
Número MDL | MFCD22380701 |
Clave InChI | VWMUCEDXVAQLNS-UHFFFAOYSA-N |
Sinónimo | bisphenol a acetate propionate, technical grade, bisphenol a acetate propionate, epoxy curative, ec-326, 4-2-4-acetyloxy phenyl propan-2-yl phenyl propanoate |
PubChem CID | 59317407 |
Nombre IUPAC | [4-[2-(4-acetiloxifenil)propan-2-il]fenil] propanoato |
SMILES | CCC(=O)OC1=CC=C(C=C1)C(C)(C)C2=CC=C(C=C2)OC(=O)C |
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
- Recommended for use as an epoxy curative to improve toughness, hydrophobicity, and hydrolytic stability
Identificadores de productos químicos
1071466-57-3 | |
326.392 | |
VWMUCEDXVAQLNS-UHFFFAOYSA-N | |
59317407 | |
CCC(=O)OC1=CC=C(C=C1)C(C)(C)C2=CC=C(C=C2)OC(=O)C |
C20H22O4 | |
MFCD22380701 | |
bisphenol a acetate propionate, technical grade, bisphenol a acetate propionate, epoxy curative, ec-326, 4-2-4-acetyloxy phenyl propan-2-yl phenyl propanoate | |
[4-[2-(4-acetiloxifenil)propan-2-il]fenil] propanoato |
Especificaciones
Bisphenol A acetate propionate | |
Epoxy curative, EC-326 | |
Blanco a amarillo sólido | |
MFCD22380701 | |
VWMUCEDXVAQLNS-UHFFFAOYSA-N | |
[4-[2-(4-acetiloxifenil)propan-2-il]fenil] propanoato | |
59317407 |
Blanco a amarillo | |
1071466-57-3 | |
C20H22O4 | |
bisphenol a acetate propionate, technical grade, bisphenol a acetate propionate, epoxy curative, ec-326, 4-2-4-acetyloxy phenyl propan-2-yl phenyl propanoate | |
CCC(=O)OC1=CC=C(C=C1)C(C)(C)C2=CC=C(C=C2)OC(=O)C | |
326.392 | |
326.39 |
Seguridad y manipulación
Declaración de GHS H
H315-H319-H335
Provoca irritación cutánea.
Provoca irritación ocular grave.
Puede irritar las vías respiratorias.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335.
missing translation for 'tsca' : No
Recommended Storage : Mantener en frío