Learn More
mb2 Asientos Boeing negros
Click to view available options
:
Capacidad: 5 ruedas
Description
- Chair in artificial leather
- Seat and back in black artificial leather upholstered
- Backrest mechanism in height, depth and permanent contact adjustable and in every position lockable
- Pneumatic cylinder mid
- 5-spoke base injected in nylon and fiberglass reinforced by a galvanized steel ring.
- Hard floor castors.
Specifications
Specifications
| Tipo de producto | Asiento Boeing |
| Color | Negro |
| Ajustable (altura) | Yes |
| Ajustables | Height |
| Estilo de la parte posterior | Ergonomic |
| Altura (métrico) | 700 mm |
| Admite | 5 ruedas |
| Cantidad | 1 unidad |
| Altura del asiento | 490 to 690 mm |
| Con reposabrazos | No |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction