Descripción
Safely transfer chemicals and caustic solutions including disinfectants, soap, and ink. Won’t crack or weaken—ensure longer pump life in demanding applications. Engineered to resist abrasion and weathering. E in., cellent fle in., ural fatigue and abrasion resistance. Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selection
Especificaciones
Especificaciones
| Diámetro interior (imperial) | 3/16 in |
| Material | TPE |
| Diámetro exterior (imperial) | 5/16 in |
| Longitud global (imperial) | 25 ft. |
| Método de esterilización | No |
| Color | Negro |