Learn More
Cole-Parmer Black TPE Tubing
Código de producto. 30053997 Tienda Cole-Parmer Productos
Change view
Click to view available options
Diámetro interior (imperial):
1/2 in
1/4 in
1/8 in
3/16 in
3/8 in
5/8 in
Diámetro exterior (imperial):
1/2 in
1/4 in
13/16 in
3/4 in
3/8 in
5/16 in
5/8 in
7/16 in
Tamaño de la unidad:
7.60 metro
Código de producto. Diámetro interior (imperial) Diámetro exterior (imperial) unitSize
30053997 1/4 in 3/8 in 7.60 metro
30053260 3/16 in 3/8 in 7.60 metro
30053804 1/4 in 1/2 in 7.60 metro
30053044 1/2 in 3/4 in 7.60 metro
30053088 3/16 in 5/16 in 7.60 metro
30054240 5/8 in 13/16 in 7.60 metro
30053390 1/4 in 7/16 in 7.60 metro
30054068 1/8 in 3/8 in 7.60 metro
30054375 1/8 in 1/4 in 7.60 metro
30053919 3/8 in 1/2 in 7.60 metro
30053147 3/8 in 5/8 in 7.60 metro
30054277 1/2 in 5/8 in 7.60 metro
12 options
Código de producto. 30053997
Marca: Cole-Parmer 5012125
Solo quedan null
Añadir a la cesta
Añadir a la cesta
Resistant to light and ozone for longer pumping life
Safely transfer chemicals and caustic solutions including disinfectants, soap, and ink. Won’t crack or weaken—ensure longer pump life in demanding applications. Engineered to resist abrasion and weathering. E in., cellent fle in., ural fatigue and abrasion resistance. Ideal for general-purpose, laboratory, and single-use applications. Free technical application support available to assist with tubing selection
Especificaciones
Diámetro interior (imperial) 1/4 in
Material TPE
Diámetro exterior (imperial) 3/8 in
Longitud global (imperial) 25 ft.
Método de esterilización Yes
Color Negro