missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SHOWA™ Blue Chemical Protection Gloves, Model CS700
Click to view available options
Tamaño:
7
8
9
10
11
Descripción
- A double-dipped nitrile coating provides chemical protection and ensures the gloves are highly durable to keep the wearer safe from harmful substances.
- The combination of a seamless liner and natural latex-free design is kind to the skin.
Especificaciones
Chlorinated | No |
Color | Blue |
Finger Thickness (English) | 45.6 mil |
Finger Thickness (Metric) | 1.16 mm |
Latex Free | Yes |
Material | Nitrile, Polyester |
Palm Thickness (English) | 45.6 mil |
Palm Thickness (Metric) | 1.16 mm |
Tamaño | 7 |
Grosor (imperial) | 45.6 mil |
Mostrar más |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'