missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMAL1 Monoclonal Antibody (1C5), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA515624
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
MA5-15624 targets ARNTL in indirect ELISA, IHC and WB applications and shows reactivity with Human samples. The MA5-15624 immunogen is purified recombinant fragment of human ARNTL expressed in E. Coli.<br>. MA5-15624 detects ARNTL which has a predicted molecular weight of approximately 68.7kDa.
The protein encoded by this gene is a basic helix-loop-helix protein that forms a heterodimer with CLOCK. This complex binds an E-box upstream of the PER1 gene, activating this gene and possibly other circadian rhythym-associated genes. Three transcript variants encoding two different isoforms have been found for this gene.Especificaciones
BMAL1 | |
Monoclonal | |
Conc. Not Determined | |
ascites with 0.03% sodium azide | |
O00327, Q9WTL8 | |
Arntl | |
Purified recombinant fragment of human ARNTL expressed in E. Coli. | |
RUO | |
11865, 406 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ELISA, Immunohistochemistry, Western Blot | |
1C5 | |
Unconjugated | |
Arntl | |
Arnt3; ARNTL; ARNT-like protein 1, brain and muscle; aryl hydrocarbon receptor nuclear translocator like; aryl hydrocarbon receptor nuclear translocator-like; aryl hydrocarbon receptor nuclear translocator-like protein 1; basic-helix-loop-helix-PAS orphan MOP3; basic-helix-loop-helix-PAS protein MOP3; bHLHe5; bHLH-PAS protein JAP3; BMAL1; Bmal1 protein; BMAL1b; bmal1b'; BMAL1c; Brain and muscle ARNT-like 1; brain and muscle Arnt-like protein 1; CG8727 PA; class E basic helix-loop-helix protein 5; cycle; JAP3; LOW QUALITY PROTEIN: aryl hydrocarbon receptor nuclear translocator-like protein 1; member of PAS protein 3; member of PAS superfamily 3; MGC47515; MOP3; PAS domain-containing protein 3; PASD3; testis tissue sperm-binding protein Li 50e; tic | |
Mouse | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 |