missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BMP4 Mouse anti-Human, Clone: OTI1D1, liquid, TrueMAB™
Mouse Monoclonal Antibody
Marca: Origene Technologies TA803611
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
The protein encoded by this gene is a member of the bone morphogenetic protein family which is part of the transforming growth factor-beta superfamily. The superfamily includes large families of growth and differentiation factors. Bone morphogenetic proteins were originally identified by an ability of demineralized bone extract to induce endochondral osteogenesis in vivo in an extraskeletal site. This particular family member plays an important role in the onset of endochondral bone formation in humans, and a reduction in expression has been associated with a variety of bone diseases, including the heritable disorder Fibrodysplasia Ossificans Progressiva. Alternative splicing in the 5′ untranslated region of this gene has been described and three variants are described, all encoding an identical protein.Especificaciones
BMP4 | |
Monoclonal | |
1 mg/mL | |
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
P12644 | |
BMP4 | |
Human recombit protein fragment corresponding to amino acids 293-408 of human BMP4 produced in E.coli. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG2b |
Western Blot | |
OTI1D1 | |
Unconjugated | |
BMP4 | |
BMP2B, BMP2B1, MCOPS6, OFC11, ZYME | |
Mouse | |
Affinity Chromatography | |
RUO | |
652 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Corrección del contenido de un producto
El Programa Encompass de Fisher Scientific ofrece artículos que no forman parte de nuestra cartera de distribución. Estos productos no suelen tener imágenes ni descripciones detalladas. Sin embargo, nos comprometemos a mejorar su experiencia de compra. Utilice el siguiente formulario para hacernos llegar sus comentarios sobre el contenido de este producto.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido