missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BOLL Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA582633
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Immunogen sequence: ATTQYLPGQW QWSVPQPSAS SAPFLYLQPS EVIYQPVEIA QDGGCVPPPL SLMETSVPEP YSDHGVQATY HQVYAPSAI
This gene belongs to the DAZ gene family required for germ cell development. It encodes an RNA-binding protein which is more similar to Drosophila Boule than to human proteins encoded by genes DAZ (deleted in azoospermia) or DAZL (deleted in azoospermia-like). Loss of this gene function results in the absence of sperm in semen (azoospermia). Histological studies demonstrated that the primary defect is at the meiotic G2/M transition. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.Especificaciones
BOLL | |
Polyclonal | |
Unconjugated | |
BOLL | |
4930554P13Rik; 4930597B14Rik; bol, boule-like; bol, boule-like (Drosophila); Boll; BOULE; boule homolog, RNA binding protein; boule-like RNA binding protein; boule-like RNA-binding protein; protein boule-like; RGD1559527 | |
Rabbit | |
Anitgen affinity chromatography, Antigen affinity chromatography | |
RUO | |
66037 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8N9W6 | |
BOLL | |
Recombinant protein corresponding to Human BOLL | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido