missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BOLL Polyclonal Antibody, Invitrogen™
Goat Polyclonal Antibody
Marca: Invitrogen PA5142068
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Additional Information: This antibody is expected to recognize both reported isoforms (NP_932074.1; NP_149019.1). This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
This gene belongs to the DAZ gene family required for germ cell development. It encodes an RNA-binding protein which is more similar to Drosophila Boule than to human proteins encoded by genes DAZ (deleted in azoospermia) or DAZL (deleted in azoospermia-like). Loss of this gene function results in the absence of sperm in semen (azoospermia). Histological studies demonstrated that the primary defect is at the meiotic G2/M transition. Two alternatively spliced transcript variants encoding distinct isoforms have been found for this gene.Especificaciones
BOLL | |
Polyclonal | |
Unconjugated | |
BOLL | |
4930554P13Rik; 4930597B14Rik; bol, boule-like; bol, boule-like (Drosophila); Boll; BOULE; boule homolog, RNA binding protein; boule-like RNA binding protein; boule-like RNA-binding protein; protein boule-like; RGD1559527 | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
66037 | |
-20° C, Avoid Freeze/Thaw Cycles | |
IgG |
Immunohistochemistry (Paraffin), Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q8N9W6 | |
BOLL | |
Peptide with sequence C-QPEPIKTVWSIHY. | |
100 μg | |
Primary | |
Human | |
Liquid |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido