Learn More
Invitrogen™ Bovine Rhodopsin (aa 338-348) Synthetic Peptide
Synthetic Peptide
Marca: Invitrogen™ PEP174
Detalles adicionales : CAS : 69-74-9 Peso : 0.01000kg
Descripción
Bovine Rhodopsin (residues 338-348) Synthetic Peptide, PEP-174, from Invitrogen.
PEP-174 is a 12 amino acid synthetic peptide whose sequence is derived from bovine rhodopsin protein. The sequence of this peptide is (amino to carboxy terminus): C-S (338)-K-T-E-T-S-Q-V-A-P-A (348)This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and bovine rhodopsin, catalog # PA1-729. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.Especificaciones
509933 | |
purified | |
Bovine Rhodopsin (aa 338-348) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Bovine | |
P02699 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
RHO | |
No preservative | |
50 μg | |
C-S(338)-K-T-E-T-S-Q-V-A-P-A(348) |