missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Bovine Rotavirus Antigen ELISA Kit (Colorimetric)
High sensitivity and specificity for detection of bovine rotavirus antigen.
Marca: Novus Biologicals™ NBP2-60130-1KIT
Detalles adicionales : Peso : 0.00970kg
Descripción
The Bovine Rotavirus Antigen ELISA Kit (Colorimetric) measures Bovine Rotavirus Antigen in feces.Especificaciones
Qualitative | |
Fecal Extract | |
Bovine | |
Instruction manual, Positive Control, Negative Control, Stop Solution, Wash Buffer, Substrate B, Substrate A, HRP-conjugate, Assay plate, Adhesive Strip (For 96 wells) | |
RUO | |
50 to 100 μL | |
Rotavirus Antigen |
ELISA Kit (Colorimetric) | |
For detection of bovine rotavirus antigen | |
ELISA | |
1 Kit | |
Cell Biology | |
Store the unopened product at 2° to 8°C. Protect from light. Do not use past expiration date. |
For Research Use Only.