missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTNL3 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA524387
Detalles adicionales : Peso : 0.01000kg
Descripción
BTNL3 is expressed in small intestine, colon, testis, spleen, and leukocyte and plays a role in lipid metabolism. Two isoforms have been described.Especificaciones
BTNL3 | |
Polyclonal | |
Unconjugated | |
BTNL3 | |
BTN9.1; BTNL3; BTNLR; butyrophilin like 3; butyrophilin-like 3; butyrophilin-like protein 3; Butyrophilin-like receptor; COLF4100; UNQ744/PRO1472 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
10917 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Flow Cytometry, Western Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
Q6UXE8 | |
BTNL3 | |
KLH conjugated synthetic peptide between 427-455 amino acids from the C-terminal region of human BTNL3 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |