missing translation for 'onlineSavingsMsg'
Learn More
Learn More
c-Myc Antibody (9E11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Marca: Novus Biologicals NB200-108
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
c-Myc Monoclonal specifically detects c-Myc in Human, Mouse, Rat, Chicken, Yeast samples. It is validated for Western Blot, Chromatin Immunoprecipitation, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Flow (Intracellular), CyTOF-ready.
Especificaciones
c-Myc | |
Monoclonal | |
1.0 mg/mL | |
Western Blot 1:500-1:1000, Flow Cytometry 1:200-1:400, ELISA 1:100-1:2000, Immunohistochemistry 1:100, Immunocytochemistry/Immunofluorescence reported in scientific literature (PMID 33542232), Immunoprecipitation 2ug/mg lysate, Immunohistochemistry-Paraffin 1:100, Immunohistochemistry-Frozen 1:100, Flow (Intracellular), Chromatin Immunoprecipitation (ChIP) 2 ug/ 500 ug extract, CyTOF-ready | |
avian myelocytomatosis viral oncogene homolog, BHLHE39, bHLHe39MRTL, Class E basic helix-loop-helix protein 39, c-Myc, MYC, myc proto-oncogene protein, MYCC, myc-related translation/localization regulatory factor, Proto-oncogene c-Myc, Transcription factor p64, v-myc avian myelocytomatosis viral oncogene homolog, v-myc myelocytomatosis viral oncogene homolog (avian) | |
Mouse | |
48.8 kDa | |
0.1 mL | |
Autophagy, Cancer, Cancer Stem Cells, Cell Cycle and Replication, Chromatin Research, Core ESC Like Genes, Epigenetics, Epitope Tags, Stem Cell Markers, Transcription Factors and Regulators, Tumor Suppressors | |
4609 | |
Human, Mouse, Chicken, Yeast | |
Purified |
Western Blot, Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry | |
900000000000 | |
Unconjugated | |
P01106 | |
MYC | |
A synthetic peptide corresponding to amino acids 408-420 (AEEQKLISEEDL) of human c-Myc Antibody (9E11), conjugated to KLH. [UniProt# P01106] | |
Protein A or G purified | |
RUO | |
Primary | |
Human, mouse, Saccharomyces and chicken. | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2a κ |
Ofertas especiales
-
R&D Systems
,
Bio-Techne
Impulse una nueva temporada de crecimiento y descubrimiento
Ahorre un 20 % en proteínas, anticuerpos, ELISAs y ensayos LuminexPromo Code: 25306Expires: 2025-03-23
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
c-Myc Antibody (9E11) - BSA Free, Novus Biologicals™ > 0.1mL; Unlabeled
For Research Use Only
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido