Learn More
CA125 Monoclonal Antibody (Ov185:1), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA511579
Detalles adicionales : Peso : 0.01000kg
Descripción
MA5-11579 targets CA-125 in IHC (P) applications and shows reactivity with Human samples. The MA5-11579 immunogen is partially purified mucin fraction from a pool of cancer tissue from patients with epithelial ovarian cancer.
CA 125 determinant is present on a high molecular weight, mucin like glycoprotein of high molecular weight. CA 125 has been found on frozen sections of amnion and derivatives of coelomic and mullerian epithelium, including pleura, pericardium and peritoneum. In adult tissues, epithelial cells of fallopian tube, endometrium and endocervix; pancreas, colon, gall bladder, stomach, kidney, apocrine sweat gland and mammary gland. It is also found in mesothelial cell lining of pleura, pericardium and peritoneum. It is found in ovarian tumors of serous, endometrioid or clear cell types and adenocarcinomas of mullerian type.Especificaciones
CA125 | |
Monoclonal | |
Conc. Not Determined | |
tissue culture supernatant with 0.09% sodium azide | |
Q8WXI7 | |
MUC16 | |
Partially purified mucin fraction from a pool of cancer tissue from patients with epithelial ovarian cancer | |
RUO | |
94025 | |
4° C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
Ov185:1 | |
Unconjugated | |
MUC16 | |
1110008I14Rik; Ca125; CA-125; CA125 ovarian cancer antigen; ca-ovarian; FLJ14303; Gm1480; MUC16; MUC-16; mucin 16; mucin 16, cell surface associated; mucin-16; Ovarian cancer-related tumor marker CA125; ovarian carcinoma antigen CA125; RGD1308422 | |
Mouse | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |