Learn More
CA150 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA540604
Detalles adicionales : Peso : 0.00010kg
Descripción
Peptide sequence: LLEREEKEKL FNEHIEALTK KKREHFRQLL DETSAITLTS TWKEVKKIIK Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Yeast: 83%; Zebrafish: 86%
CA150 Polyclonal antibody specifically detects CA150 in Human samples. It is validated for Western Blot.Especificaciones
CA150 | |
Polyclonal | |
Unconjugated | |
TCERG1 | |
2410022J09Rik; 2900090C16Rik; AI428505; CA150; CA150b; coactivator of 150 kD; co-activator of 150 kDa; FBP 28; FBP23; FBP28; Formin-binding protein 28; LOW QUALITY PROTEIN: transcription elongation regulator 1; MGC133200; p144; Taf2s; TATA box binding protein (TBP)-associated factor, RNA polymerase II, S, 150kD; TATA box-binding protein-associated factor 2S; TCERG1; Transcription elongation regulator 1; transcription elongation regulator 1 (CA150); Transcription elongation regulator 1-like protein; transcription factor CA150; transcription factor CA150b; Urn1 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
10915 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O14776 | |
TCERG1 | |
Synthetic peptide directed towards the middle region of human TCERG1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |