missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Calceína, MP Biomedicals™
Click to view available options
Cantidad:
10 g
25 g
Especificaciones
Nombre del producto químico o material | Calcein |
Nombre de nota | Suitable for fluorometric determination of calcium and EDTA titration of calcium in the presence of magnesium. |
Formula Weight (peso de la fórmula) | 622.5 |
Color | Naranja |
Cantidad | 10 g |
Forma física | Polvo |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'