missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase IV Monoclonal Antibody (4G6), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA518513
Detalles adicionales : Peso : 0.00010kg
Descripción
Peptide Sequence: VQAESSNYPC LVPVKWGGNC QKDRQSPINI VTTKAKVDKK LGRFFFSGYD KKQTWTVQNN GHSVMMLLEN KASISGGGLP APYQAKQLHL HWSDLPYKGS
Carbonic Anhydrase IV Monoclonal antibody specifically detects Carbonic Anhydrase IV in Human samples. It is validated for ELISA, Western Blot.Especificaciones
Carbonic Anhydrase IV | |
Monoclonal | |
0.2-1.0 mg/mL | |
PBS with no preservative; pH 7.4 | |
P22748 | |
CA4 | |
CA4 (NP_000708.1, 27 a.a. ∽ 126 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
ELISA, Western Blot | |
4G6 | |
Unconjugated | |
CA4 | |
AW456718; CA IV; CA4; CAIV; CA-IV; Car4; Carbonate dehydratase IV; carbonic anhydrase 4; Carbonic anhydrase IV; carbonic dehydratase IV; RP17 | |
Mouse | |
Affinity chromatography | |
RUO | |
762 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |