missing translation for 'onlineSavingsMsg'
Learn More

Carbonic Anhydrase VA Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Marca:  Invitrogen PA539078

Detalles adicionales : Peso : 0.00900kg

Código de producto. 15525991

  • 472.00€ / 100 microgramos
Fecha estimada de envío: 03-09-2024
para ver el stock

Descripción

Descripción

Carbonic anhydrases (CAs) are members of a large family of zinc metalloenzymes that catalyze the reversible hydration of carbon dioxide. CAs are involved in a variety of biological processes, including respiration, calcification, acid-base balance, bone resorption, and the formation of aqueous humor, cerebrospinal fluid, saliva, and gastric juice. They show extensive diversity in distribution and in their subcellular localization. The human CA5A gene encodes for the CA V protein which is expressed in the mitochondrial matrix in liver tissues, but can also be detected in many other tissues. In ureagenesis, the CA V protein may be involved in providing bicarbonate to carbamoyl phosphate synthetase.
Especificaciones

Especificaciones

Carbonic Anhydrase VA
Polyclonal
Unconjugated
CA5A
Carbonic anhydrase 5A, mitochondrial, Carbonate dehydratase VA, Carbonic anhydrase VA, CA-VA, CA5A, CA5
Rabbit
Antigen affinity chromatography
RUO
763
-20°C
Liquid
Western Blot
1 mg/mL
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4
P35218
CA5A
A synthetic peptide derived from the internal region of human CA5A
100 μg
Primary
Human
Antibody
IgG
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales