missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Carbonic Anhydrase VA Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA536931
Detalles adicionales : Peso : 0.00910kg
Descripción
This antibody detects endogenous protein at a molecular weight of 35 kDa. Purity is >95% by SDS-PAGE.
Carbonic Anhydrase VA Polyclonal antibody specifically detects Carbonic Anhydrase VA in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Western Blot.Especificaciones
Carbonic Anhydrase VA | |
Polyclonal | |
Unconjugated | |
CA5A | |
CA Y; CA5; CA5A; CA5AD; Car5; Car5a; Carbonate dehydratase VA; carbonic anhydrase 5 mitochondrial; carbonic anhydrase 5, mitochondrial; carbonic anhydrase 5A; carbonic anhydrase 5a, mitochondrial; carbonic anhydrase 5b, mitochondrial; carbonic anhydrase V, mitochondrial; carbonic anhydrase VA; carbonic anhydrase VA, mitochondrial; carbonic dehydratase; CAV; CAVA; CA-VA; GS1-21A4.1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
12352, 54233, 763 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
P23589, P35218, P43165 | |
CA5A, Car5a | |
Synthetic peptide corresponding to amino acids 223-268 of human CA5A | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |