Learn More
Carbonic Anhydrase XI Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA522339
Detalles adicionales : Peso : 0.01000kg
Descripción
Recommended positive controls: A431. Predicted reactivity: Pig (96%), Sheep (97%), Bovine (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Carbonic Anhydrase XI (CA11, CA XI) is a member of the carbonic anhydrases (CAs) family of zinc metalloenzymes that catalyze the reversible hydration of carbon dioxide. Carbonic Anhydrase XI participate in a variety of biological processes, including respiration, calcification, acid-base balance, bone resorption, and the formation of aqueous humor, cerebrospinal fluid, saliva, and gastric acid. Further, Carbonic Anhydrase XI show extensive diversity in tissue distribution and in their subcellular localization. CA XI is likely a secreted protein, however, radical changes at active site residues completely conserved in CA isozymes with catalytic activity, make it unlikely that it has carbonic anhydrase activity. It shares properties in common with two other acatalytic CA isoforms, CA VIII and CA X. CA XI is most abundantly expressed in brain, and may play a general role in the central nervous system.Especificaciones
Carbonic Anhydrase XI | |
Polyclonal | |
Unconjugated | |
CA11 | |
CA11; Car11; carbonic anhydrase 11; carbonic anhydrase XI; carbonic anhydrase-related protein 11; Carbonic anhydrase-related protein 2; carbonic anhydrase-related protein XI; carbonic anhydrase-related XI protein; CA-RP II; CARP XI; CA-RP XI; Carp2; CARP-2; CARPX1; CAR-XI; CA-XI; UNQ211/PRO237 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
770 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
0.81 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
O75493 | |
CA11 | |
Recombinant fragment corresponding to a region within amino acids 1 and 311 of CA11 (Uniprot ID#O75493) | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |