missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Catalase Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Marca: Invitrogen PA5122155
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Catalase is a homotetrameric heme-containing enzyme present within the matrix of all peroxisomes. It carries out a dismutation reaction in which hydrogen peroxide is converted to water and oxygen. Human catalase has the last four amino acids (-KANL) at the extreme C-terminus for peroxisome targeting. The monomer of human catalase is 61.3 kDa in molecular size. Catalase has been implicated as an important factor in inflammation, mutagenesis, prevention of apoptosis, and stimulation of a wide spectrum of tumors. Loss of catalase leads to the human genetic disease, acatalasemia, or Takahara's disease.Especificaciones
Catalase | |
Polyclonal | |
Unconjugated | |
Cat | |
2210418N07; Cas1; Cas-1; CAT; Cat01; catalase; catalase 1; Catl; CS1; Cs-1; CTA1; HGNC:1516; hypothetical protein LOC531682; MGC138422; MGC138424 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
847 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
P04040 | |
Cat | |
Recombinant protein Catalase. The antigen corresponds to amino acid range 401-523 of the target protein. | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |