missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CCNY Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Marca: Proteintech 18042-1-AP-150UL
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
Cyclins, such as CCNY, control cell division cycles and regulate cyclin-dependent kinases.Especificaciones
CCNY | |
Polyclonal | |
Unconjugated | |
CCNY | |
C10orf9, CBCP1, CCNY, CFP1, Cyc Y, Cyclin box protein 1, Cyclin fold protein 1, cyclin X, cyclin Y | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
219771, 361261, 67974 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q8BGU5, Q8ND76 | |
Ccny | |
CCNY Fusion Protein Ag12674 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |