missing translation for 'onlineSavingsMsg'
Learn More

Epredia™ Lab Vision™ CD10/CALLA (Neutral Endopeptidase) Ab-2, Mouse Monoclonal Antibody

Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ CD10/CALLA (Neutral Endopeptidase) Ab-2, Mouse Monoclonal Antibody.

Marca:  Epredia™ MS-728-S0

 Ver más versiones de este producto

Código de producto. 12622177

  • 165.00€ / 100 microlitros

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.

Explore más ofertas especiales
Este artículo no se puede devolver. Vea la política de devoluciones

Descripción

Descripción

CD10, also known as Common Acute Lymphocytic Leukemia Antigen (CALLA), is a cell surface enzyme with neutral metalloendopeptidase activity which inactivates a variety of biologically active peptides. CD10 is expressed on the cells of lymphoblastic, Burkitt's, and follicular germinal center lymphomas, and on cells from patients with chronic myelocytic leukemia (CML). It is also expressed on the surface of normal early lymphoid progenitor cells, immature B cells within adult bone marrow and germinal center B cells within lymphoid tissue. CD10 is also present on breast myoepithelial cells, bile canaliculi, fibroblasts, with especially high expression on the brush border of kidney and gut epithelial cells.

Host Species: Mouse

Clone: 56C6

Isotype: IgG1

Species Reactivity: Human and Rat. Others not tested.

Epitope: Extracellular

Immunogen: Recombinant external domain of CD10 protein

Molecular Weight: 100kDa

Positive Control: Small intestine

Cellular Localization: Membrane

Recommended for:

  • Immunohistochemistry (Formalin/paraffin)
Especificaciones

Especificaciones

CD10
Monoclonal
Unconjugated
Recombinant external domain of CD10 protein
IVD
Primary
Concentrated
Immunohistochemistry (Paraffin)
56C6
Mouse
100 μL
Oncogenesis
Human, Rat
IgG1
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales