Learn More
CD156c Mouse anti-Human, Brilliant Violet 650, Clone: 11G2, BD Optibuild™
Mouse Monoclonal Antibody
Marca: BD Biosciences 742790
Descripción
The 11G2 monoclonal antibody specifically binds to CD156c which is also known as Disintegrin and metalloproteinase domain-containing protein 10 (ADAM10). CD156c is a ñ70 kDa type I transmembrane glycoprotein that belongs to the ADAM family. It is widely expressed on hematopoietic and non-hematopoietic cells. CD156c is expressed on the plasma membrane, as well as by membranes within intracellular compartments and exosomes. CD156c serves as a broadly-reactive endopeptidase that cleaves membrane-bound proteins in a process known as ectodomain shedding or release. It can cleave transmembrane molecules into soluble forms and thereby regulate the functions of a variety of different receptors and ligands that are involved in cellular signaling and adhesion. These include membrane TNF, CX3CL1, CXCL16, Ephrin-A2, Notch and Delta molecules, cadherins, CD23, CD44, IL-6 Receptor/CD126, CD171, and amyloid precursor protein. CD156c expression may be constitutive or induced, such as in inflamed tissues, including arthritic joints and in the nervous system. Dysregulated CD156c expression may be associated with certain cancers and Alzheimer’s disease.
The antibody was conjugated to BD Horizon™ BV650 which is part of the BD Horizon Brilliant™ Violet family of dyes. This dye is a tandem fluorochrome of BD Horizon BV421 with an Ex Max of 405nm and an acceptor dye with an Em Max at 650nm. BD Horizon BV650 can be excited by the violet laser and detected in a filter used to detect APC-like dyes (eg, 660/20nm filter). Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. However, the spillover can be corrected through compensation as with any other dye combination. BV650 is a tandem fluorochrome of BD Horizon BV421 and an acceptor dye with an emission maximum at 650 nm. Due to the excitation and emission characteristics of the acceptor dye, there will be spillover into the APC and Alexa Fluor™ 700 detectors. BV650 will have moderate spillover into the BD Horizon™ BV711 detector.
Especificaciones
| CD156c | |
| 11G2 | |
| Brilliant Violet 650 | |
| ADAM 10, AD10, ADA10, MADM, KUZ, Kuzbanian protein homolog, HsT18717, RAK | |
| Human Jurkat Cell Line | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| Monoclonal | |
| 0.2mg/mL | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Human | |
| IgG1 κ |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.