Learn More
CD171 Monoclonal Antibody (UJ127), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA514140
Detalles adicionales : Peso : 0.01000kg
Descripción
MA5-14140 targets L1 Cell Adhesion Molecule in immunohistochemistry (paraffin), immunoprecipitation, and Western blot applications and shows reactivity with human samples. The MA5-14140 immunogen is homogenous suspension of 16 week human fetal brain. MA5-14140 has been successfully used in immunohistochemistry analysis of NCAM1 in mouse E16.5 frozen lung sections.
L1CAM/CD171 is an axonal glycoprotein belonging to the immunoglobulin supergene family. The ectodomain, consisting of several immunoglobulin-like domains and fibronectin-like repeats (type III), is linked via a single transmembrane sequence to a conserved cytoplasmic domain. This cell adhesion molecule plays an important role in nervous system development, including neuronal migration and differentiation. Mutations in the gene cause three X-linked neurological syndromes known by the acronym CRASH (corpus callosum hypoplasia, retardation, aphasia, spastic paraplegia and hydrocephalus). Alternative splicing of a neuron-specific exon is thought to be functionally relevant.Especificaciones
CD171 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.05% BSA and 0.05% sodium azide; pH 7.4 | |
P11627, P32004 | |
L1CAM | |
Homogenous suspension of 16 week human fetal brain | |
500 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG1 κ |
Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
UJ127 | |
Unconjugated | |
L1CAM | |
antigen identified by monoclonal antibody R1; CAML1; CD 171; CD171; CD171 molecule; HSAS; HSAS1; Hyd; L1; L1 cell adhesion molecule; L1CAM; MASA; MIC5; N-CAM L1; NCAML1; N-CAML1; N-CAM-L1; NCAM-L1; Nerve-growth factor-inducible large external glycoprotein; neural cell adhesion molecule L1; NILE; S10; sCD171; sL1 CAM; sL1CAM; soluble CD 171; soluble CD171; soluble L1 CAM; soluble L1CAM; SPG1 | |
Mouse | |
Protein A/G | |
RUO | |
16728, 3897 | |
4° C | |
Liquid |