missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD22 Monoclonal Antibody (eBio4KB128 (4KB128)), NovaFluor™ Blue 660-120S, eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen H053T03B08
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CD22 Monoclonal antibody specifically detects CD22 in Human samples. It is validated for Flow Cytometry.Especificaciones
CD22 | |
Monoclonal | |
0.2 μg/Test | |
PBS with 0.09% sodium azide; pH 7.2 | |
P20273 | |
Mouse | |
100 Tests | |
Primary | |
Human | |
Antibody | |
IgG1 κ |
Flow Cytometry | |
eBio4KB128 (4KB128) | |
NovaFluor Blue 660-120S | |
CD22 | |
A530093D23; B-cell receptor CD22; BL-CAM; B-lymphocyte cell adhesion molecule; B-lymphocyte cell adhesion molecule (BL-CAM); Cd22; CD22 antigen; CD22 molecule; FLJ22814; Lectin 2; Leu-14; Lyb8; Lyb-8; MGC130020; sialic acid binding Ig-like lectin 2; sialic acid-binding Ig-like lectin 2; Sialic acid-binding Ig-like lectin 2 (Siglec-2); Siglec2; Siglec-2; T-cell surface antigen Leu-14 | |
Affinity chromatography | |
RUO | |
933 | |
4° C, store in dark, DO NOT FREEZE! | |
Liquid |