Learn More
Invitrogen™ CD244 (2B4) Monoclonal Antibody (M053)

Mouse Monoclonal Antibody
Marca: Invitrogen™ MA547659
Descripción
Clone M053 mouse monoclonal antibody detects a 110 kDa* protein on SDS-PAGE Native or denatured immunoblots of human THP1 monocytes. The antibody prefers the native CD244 protein, but also detects denatured CD244. The antibody works for western blot, immunoprecipitation, immunocytochemistry, and ELISA capture.
CD244 (Natural killer (NK) cell receptor 2B4/SLAMF4) is an Ig superfamily signaling lymphocyte activation molecule (SLAM) receptor. Like all SLAM family receptors, it has an extracellular segment with two immunoglobulin (Ig)-like domains, and a cytoplasmic domain containing four immunoreceptor tyrosine-based switch motifs. CD244 does not act as a selfligand similar to other SLAM family receptors. It binds CD48, a transmembrane receptor ubiquitously expressed on hematopoietic cells. CD244 activity is controlled by the presence or absence of small cytoplasmic adapter proteins, SH2D1A/SAP and/or SH2D1B/EAT-2. Downstream signaling involves predominantly VAV1, and, to a lesser degree, INPP5D/SHIP1 and CBL. Activation of CD244 stimulates NK cell cytotoxicity, production of IFN-gamma and granule exocytosis. CD244 is involved in the regulation of CD8+ T-cell proliferation, and inhibits inflammatory responses in dendritic cells (DCs). In cancers, CD244 shows increased expression in intratumoral DCs and myeloid suppressor cells, and anti-CD244 therapies may increase infiltrating T-cells and impair tumor growth.
Especificaciones
| CD244 (2B4) | |
| Monoclonal | |
| 0.5 mg/mL | |
| PBS with 1mg/mL BSA, 50% glycerol and 0.05% sodium azide | |
| Q9BZW8 | |
| CD244 | |
| Clone (M053) was generated from a recombinant protein that included the extracellular region of human CD244 protein. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| ELISA, Immunoprecipitation, Western Blot, Immunocytochemistry | |
| M053 | |
| Unconjugated | |
| CD244 | |
| NKR2B4; SLAMF4 | |
| Mouse | |
| Protein G | |
| RUO | |
| 51744 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.