missing translation for 'onlineSavingsMsg'
Learn More

CD3e Hamster anti-Mouse, APC-Cy7, Clone: 145-2C11, BD

Hamster Monoclonal Antibody

Marca:  BD Biosciences 554843

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15887878

  • 773.00€ / cada uno
Fecha estimada de envío: 21-08-2024
para ver el stock

Descripción

Descripción

The OX-38 antibody has been reported to react with the CD4 antigen on most thymocytes, a subpopulation of mature T lymphocytes (i.e., MHC class II-restricted T cells, including most T helper cells), monocytes, macrophages, and some dendritic cells. CD4 is an antigen coreceptor on the T-cell surface which interacts with MHC class II molecules on antigen-presenting cells. It participates in T-cell activation through its association with the T-cell receptor complex and protein tyrosine kinase lck. The OX-38 antibody has been reported to bind to the same epitope of CD4 as that recognized by W3/25 mAb, which is a different epitope than that recognized by OX-35 mAb (Cat. No. 554837). In vivo blocking of some cell-mediated immune responses by mAb OX-38 has been reported. Injection of OX-38 mAb induces allograft unresponsiveness in rats, with varying results depending on the rat strain used (high or low responder). Furthermore, in vivo depletion of CD4+ lymphocytes has been reported with this antibody.

This antibody is routinely tested by flow cytometric analysis. Other applications were tested at BD Biosciences Pharmingen during antibody development only or reported in the literature.

Especificaciones

Especificaciones

CD4
Monoclonal
0.5mg/mL
Aqueous buffered solution containing ≤0.09% sodium azide.
Rat thymocyte glycoproteins
0.5 mg
Immunology
Rat
IgG2a κ
Flow Cytometry
OX-38
FITC
Mouse
Affinity Purified
RUO
Primary
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Sugerencias de productos

Sugerencias de productos

Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales

For Research Use Only.