Learn More
CD4 Rat anti-Mouse, APC, Clone: RM4-5, BD
Rat Monoclonal Antibody
185.00€ - 344.00€
Especificaciones
Antígeno | CD4 |
---|---|
Clon | RM4-5 |
Concentración | 0.2mg/mL |
Aplicaciones | Flow Cytometry |
Clasificación | Monoclonal |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15880759
|
BD Biosciences
561091 |
25 μg |
185.00€
cada uno |
Fecha estimada de envío: 21-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
15876958
|
BD Biosciences
553051 |
0.1 mg |
344.00€
cada uno |
Fecha estimada de envío: 12-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
The RM4-5 monoclonal antibody specifically binds to the CD4 (L3T4) differentiation antigen expressed on most thymocytes, subpopulations of mature T lymphocytes (i.e., MHC class II-restricted T cells, including most T helper cells and immunosuppressive regulatory T cells), and a subset of NK-T cells. CD4 has also been reported to be detected on pluripotent hematopoietic stem cells, bone marrow myeloid and B-lymphocyte precursors, intrathymic lymphoid precursors, and a subset of splenic dendritic cells. CD4 has been reported to be expressed on the plasma membrane of mouse egg cells and is involved in adhesion of the egg to MHC class II-bearing sperm. CD4 is an antigen coreceptor on the T-cell surface which interacts with MHC class II molecules on antigen-presenting cells. It participates in T-cell activation through its association with the T-cell receptor complex and protein tyrosine kinase lck. Purified RM4-5 mAb has been reported to block the binding of FITC-conjugated anti-mouse CD4 clones GK1.5 and H129.19, but not the RM4-4 clone.
Especificaciones
CD4 | |
0.2mg/mL | |
Monoclonal | |
Rat | |
Mouse | |
Mouse Thymocytes (BALB/c) | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
RM4-5 | |
Flow Cytometry | |
APC | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
IgG2a κ | |
Affinity Purified | |
Cd4; CD4 antigen; L3T4; Ly-4; T-cell surface antigen T4/Leu-3 |
For Research Use Only.