missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD43 Monoclonal Antibody (W3/13HLK), PE, Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA517386
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
MA5-17386 targets CD43/SPN in FACS applications and shows reactivity with Rat samples. The MA5-17386 immunogen is rat thymocyte membrane.
CD43 (leukosialin, sialophorin) is a transmembrane mucin-like protein with high negative charge, expressed on the surface of most hematopoietic cells. CD43 contributes to a repulsive barrier that interferes with cellular adhesion, however, in certain cases also promotes leukocyte aggregation. By interaction with actin-binding proteins ezrin and moesin CD43 plays a regulatory role in remodeling T-cell morphology and regulates cell-cell interactions during lymphocyte traffic. CD43 signaling both enhances LFA-1 adhesiveness and counteracts LFA-1 induction via other receptors. Expression of CD43 causes induction of functionally active tumor suppressor p53 protein, but in case of p53 and ARF deficiency CD43 promotes tumor proliferation and viability. CD43 appears to be an important modulator of leukocyte functions. Diseases associated with CD43 dysfunction include Wiscott-Aldrich Syndrome and Adenoid Basal Cell Carcinoma.Especificaciones
CD43 | |
Monoclonal | |
0.1 mg/mL | |
PBS with 4-5mg/mL BSA and 0.02% sodium azide | |
P13838 | |
SPN | |
Rat thymocyte membrane | |
50 μg | |
Primary | |
Rat | |
Antibody | |
IgG1 |
Flow Cytometry | |
W3/13HLK | |
PE | |
SPN | |
3E8 antigen; A630014B01Rik; B-cell differentiation antigen LP-3; CD43; CD43 cytoplasmic tail; CD43-ct; Galactoglycoprotein; GALGP; gpL115; Leukocyte sialoglycoprotein; leukosialin; leukosianin; LOC101120462; LOW QUALITY PROTEIN: leukosialin; Lsn; Lsn1; Ly48; Ly-48; Lymphocyte antigen 48; sialophorin; sialophorin (gpL115, leukosialin, CD43); sialophorin gpL115; SPN; W3/13 antigen | |
Mouse | |
Protein G | |
RUO | |
24796 | |
4° C, do not freeze | |
Liquid |