Learn More
CD45.2 Mouse anti-Mouse, FITC, Clone: 104, BD
Mouse Monoclonal Antibody
221.00€ - 510.00€
Especificaciones
Antígeno | CD45.2 |
---|---|
Clon | 104 |
Concentración | 0.5mg/mL |
Aplicaciones | Flow Cytometry |
Clasificación | Monoclonal |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15891419
|
BD Biosciences
561874 |
0.1 mg |
221.00€
cada uno |
Fecha estimada de envío: 21-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
15897378
|
BD Biosciences
553772 |
0.5 mg |
510.00€
cada uno |
Fecha estimada de envío: 21-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
The 104 clone has been reported to react with CD45 (Leukocyte Common Antigen) on all leukocytes of most mouse strains (eg, A, AKR, BALB/c, CBA/Ca, CBA/J, C3H/He, C57BL, C57BR, C57L, C58, DBA/1, DBA/2, NZB, SWR, 129). This alloantigen was originally named Ly-5.1, and this was the designation at the time that the antibody was characterized. The designation was later changed from Ly-5.1 to Ly-5.2 to conform with the convention that the .2 alloantigen designations be assigned to the C57BL/6 strain. mAb 104 has been reported not to react with leukocytes of the mouse strains expressing the CD45.1 alloantigen (eg, RIII, SJL/J, STS/A, and DA). CD45 is a member of the Protein Tyrosine Phosphatase (PTP) family: its intracellular (COOH-terminal) region contains two PTP catalytic domains, and the extracellular region is highly variable due to alternative splicing of exons 4, 5, and 6 (designated A, B, and C, respectively), plus differing levels of glycosylation. The CD45 isoforms detected in the mouse are cell type-, maturation-, and activation state-specific. The CD45 isoforms play complex roles in T-cell and B-cell antigen receptor signal transduction. The 104 antibody has been reported to inhibit some responses of B cells, from mice expressing the CD45.2 alloantigen, to certain antigens and LPS. In addition, reduction of serum IgG levels and amelioration of autoimmune renal pathology were reported in mAb 104-treated systemic lupus erythematosus-prone mice.
Especificaciones
CD45.2 | |
0.5mg/mL | |
Monoclonal | |
Mouse | |
Mouse | |
B10.S mouse thymocytes and splenocytes | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
104 | |
Flow Cytometry | |
FITC | |
RUO | |
Aqueous buffered solution containing protein stabilizer and ≤0.09% sodium azide. | |
IgG2a κ | |
Affinity Purified | |
Ly-5.2; T200; LCA; Leukocyte common antigen; Ptprc |
For Research Use Only.