Learn More
CD45R/B220 Rat anti-Mouse, PerCP-Cy5.5, Clone: RA3-6B2, BD
Rat Monoclonal Antibody
199.00€ - 570.00€
Especificaciones
Antígeno | CD45R |
---|---|
Clon | RA3-6B2 |
Concentración | 0.2mg/mL |
Aplicaciones | Flow Cytometry |
Clasificación | Monoclonal |
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
---|---|---|---|---|---|---|---|---|---|
Código de producto | Marca | Cantidad | Precio | Cantidad y disponibilidad | |||||
15880769
|
BD Biosciences
561101 |
25 μg |
199.00€
cada uno |
Fecha estimada de envío: 21-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
15876848
|
BD Biosciences
552771 |
0.1 mg |
570.00€
cada uno |
Fecha estimada de envío: 21-08-2024 Inicie sesión para ver el stock |
Por favor, inicie sesión para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo. | ||||
Descripción
The rat anti-mouse CD45R antibody (clone RA3-6B2) has been reported to react with an epitope on the extracellular domain of the transmembrane CD45 glycoprotein which is dependent upon the expression of exon A and specific carbohydrate residues. It is expressed on B lymphocytes at all stages from pro-B through mature and activated B cell, but it is decreased on plasma cells and a subset of memory B cells. The levels of CD45R expression on the B-cell lineage appear to be developmentally regulated. It is also reportedly found on the abnormal T cells involved in the lymphadenopathy of lpr/lpr and gld/gld mutant mice, on lytically active subsets of lymphokine-activated killer cells (NK cells and non-MHC-restricted CTL), on apoptotic T lymphocytes of mice injected with bacterial superantigen, on a population of NK-cell precursors in the bone marrow, and on B-lymphocyte, T-lymphocyte, and macrophage progenitors in fetal liver. The CD45R antigen has been reported not to be on hematopoietic stem cells, naive T lymphocytes, or MHC-restricted CTL. CD45 is a member of the Protein Tyrosine Phosphatase (PTP) family: Its intracellular (COOH-terminal) region contains two PTP catalytic domains, and the extracellular region is highly variable due to alternative splicing of exons 4, 5, and 6 (designated A, B, and C, respectively), plus differing levels of glycosylation. The CD45 isoforms detected in the mouse are cell type-, maturation, and activation state-specific. The CD45 isoforms play complex roles in T-cell and B-cell antigen receptor signal transduction. CD45R is commonly used as a pan B-cell marker; however, CD19 expression, detectable by the rat anti-mouse CD19 antibody (clone 1D3), is reported to be more restricted to the B-cell lineage. The rat anti-mouse CD45R antibody (clone RA3-6B2) has been reported to enhance isotype switching during in vitro B-cell responses and to inhibit in vivo B-cell responses. Cross-reaction of the RA3-6B2 clone with activated human T lymphocytes has also been reportedly observed.
Especificaciones
CD45R | |
0.2mg/mL | |
Monoclonal | |
Rat | |
Human, Mouse | |
Mouse Abelson Leukemia Virus-Induced pre-B tumor cells | |
Primary | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
RA3-6B2 | |
Flow Cytometry | |
PerCP-Cyanine5.5 | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
IgG2a κ | |
Affinity Purified |
For Research Use Only.