missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD5 Mouse anti-Human, BUV661, Clone: UCHT2, BD Biosciences™
Mouse Monoclonal Antibody
Marca: BD Biosciences 741598
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
CD5 Monoclonal antibody specifically detects CD5 in Human samples. It is validated for Flow Cytometry
Especificaciones
| CD5 | |
| Monoclonal | |
| Brilliant Ultraviolet 661 | |
| CD5 antigen (p56-62); T1; Tp67; LEU1; Lymphocyte antigen T1/Leu-1 | |
| Human T cells | |
| RUO | |
| Human | |
| IgG1 κ |
| Flow Cytometry | |
| UCHT2 | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| Mouse | |
| 50 μg | |
| Primary | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon BUV661 under optimal conditions that minimize unconjugated dye and antibody. |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering