missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CD57 Monoclonal Antibody (HNK-1 (Leu-7)), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA511605
Este artículo no se puede devolver.
Vea la política de devoluciones
Descripción
MA5-11605 targets CD57 in FACS, WB and IHC (P) applications and shows reactivity with Human samples. The MA5-11605 immunogen is membrane extracts of the HSB-2 T lymphoblastoid cell line.
CD57, also known as HNK-1 and Leu-7, is an oligosaccharide antigenic determinant present on a variety of polypeptides, lipids, and chondroitin sulphate proteoglycans. Its function is poorly understood. CD57 is expressed on a subset of natural killer (NK) cells and NK T cells.Especificaciones
CD57 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
Q9P2W7 | |
B3GAT1 | |
Membrane extracts of the HSB-2 T lymphoblastoid cell line | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgM |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
HNK-1 (Leu-7) | |
Unconjugated | |
B3GAT1 | |
HNK-1; Leu-7 | |
Mouse | |
Ammonium sulfate precipitation | |
RUO | |
27087 | |
4° C | |
Liquid |