missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cdc7 Monoclonal Antibody (DCS-341), Invitrogen™
Mouse Monoclonal Antibody
Marca: Invitrogen MA512589
Detalles adicionales : Peso : 0.01000kg
Descripción
MA5-12589 targets CDC7 Kinase in IHC (P), IP, and WB applications and shows reactivity with Human samples. The MA5-12589 immunogen is gST fusion protein full length CDC7.
CDC7 is a cell division cycle 7 homolog protein that is critical for the G1/S transition and is also essential for initiation of DNA replication as cell division occurs. CDC7 is expressed in many normal tissues, but the overexpression of CDC7 may be associated with neoplastic transformation for some tumors and transformed cell lines. CDC7/DBF4 kinase promotes S phase by alleviating an inhibitory activity in Mcm4 that evolved to integrate several protein kinase. CDC7 encodes a cell division cycle protein with kinase activity that is critical for the G1/S transition. The yeast homolog is also essential for initiation of DNA replication as cell division occurs. Overexpression of this gene product may be associated with neoplastic transformation for some tumors. Multiple alternatively spliced transcript variants that encode the same protein have been detected.Especificaciones
Cdc7 | |
Monoclonal | |
0.2 mg/mL | |
PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4 | |
O00311 | |
CDC7 | |
GST fusion protein full length CDC7 | |
500 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
DCS-341 | |
Unconjugated | |
CDC7 | |
AI597260; Cdc7; CDC7 (cell division cycle 7, S. cerevisiae, homolog)-like 1; Cdc7 Kinase; Cdc7l1; CDC7-like 1; CDC7-related kinase; cell division cycle 7; cell division cycle 7 (S. cerevisiae); cell division cycle 7 homolog; cell division cycle 7-like 1; cell division cycle 7-like protein 1; cell division cycle 7-related protein kinase; HsCDC7; Hsk1; huCDC7; MGC117361; MGC126237; MGC126238; MGC91067; muCdc7 | |
Mouse | |
Protein G | |
RUO | |
8317 | |
4° C | |
Liquid |